Tisk…

Isopropyl methanesulphonate 97%

Dodavatel: Apollo Scientific

OR932406-10G OR932406-25G OR932406-100G
APOSOR932406-10GEA 5200 CZK
APOSOR932406-10G APOSOR932406-25G APOSOR932406-100G
Isopropyl methanesulphonate 97%
Isopropyl methanesulphonate
Vzorec: C₄H₁₀O₃S
MW: 138,181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047802
CAS číslo: 926-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR