4-(2-Methoxyethyl)piperidine hydrochloride 95%
Dodavatel: Apollo Scientific
OR932381-1G
OR932381-250MG
APOSOR932381-1GEA
3060
CZK
APOSOR932381-1G
APOSOR932381-250MG
4-(2-Methoxyethyl)piperidine hydrochloride 95%
4-(2-Methoxyethyl)piperidine hydrochloride
Vzorec:
C₈H₁₈ClNO Storage Temperature: Ambient |
MDL Number:
MFCD10687230 CAS číslo: 868849-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...