Tisk…

Isopropyl salicylate 95%

Dodavatel: Apollo Scientific

OR932317-100G OR932317-25G
APOSOR932317-100GEA 5130 CZK
APOSOR932317-100G APOSOR932317-25G
Isopropyl salicylate 95%
Isopropyl salicylate
Vzorec: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00035703
CAS číslo: 607-85-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR