4,6-Dichloro-2-cyclopropylpyrimidine 98%
Dodavatel: Apollo Scientific
OR931925-1G
OR931925-5G
APOSOR931925-1GEA
2860
CZK
APOSOR931925-1G
APOSOR931925-5G
4,6-Dichloro-2-cyclopropylpyrimidine 98%
4,6-Dichloro-2-cyclopropylpyrimidine
Vzorec:
C₇H₆Cl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD11046809 CAS číslo: 7043-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...