Tisk…

H-Phe(2,4-Dime)-OH 97%

Dodavatel: Apollo Scientific

Synonyms: H-Phe(2.4-Dimethyl)-OH

OR931727-1G OR931727-500MG
APOSOR931727-1GEA 12470 CZK
APOSOR931727-1G APOSOR931727-500MG
H-Phe(2,4-Dime)-OH 97%
H-Phe(2,4-Dime)-OH
Vzorec: C₁₁H₁₅NO₂
MW: 193,2 g/mol
Storage Temperature: Ambient
CAS číslo: 259726-56-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR