Tisk…

(±)-trans-1,2-Cyclopentanediol 98%

Dodavatel: Apollo Scientific

Synonyms: (±)-trans-Cyclopentane-1,2-diol, trans-Cyclopentane-1,2-diol

OR931586-25G OR931586-5G
APOSOR931586-25GEA 8830 CZK
APOSOR931586-25G APOSOR931586-5G
(±)-trans-1,2-Cyclopentanediol 98%
(±)-trans-1,2-Cyclopentanediol
Vzorec: C₅H₁₀O₂
MW: 102,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082582
CAS číslo: 5057-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR