3-Hydroxyphenylacetylene 96%
Dodavatel: Apollo Scientific
OR931307-1G
OR931307-5G
APOSOR931307-1GEA
612
CZK
APOSOR931307-1G
APOSOR931307-5G
3-Hydroxyphenylacetylene 96%
3-Hydroxyphenylacetylene
Vzorec:
C₈H₆O Storage Temperature: Ambient |
MDL Number:
MFCD00078347 CAS číslo: 10401-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...