Tisk…

2-Propoxyphenol 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Hydroxy-2-propoxybenzene

OR931281-5G
APOSOR931281-5GEA 1820 CZK
APOSOR931281-5G
2-Propoxyphenol 95%
2-Propoxyphenol
Vzorec: C₉H₁₂O₂
MW: 152,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655033
CAS číslo: 6280-96-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR