Tisk…

(±)-2,2-Dimethyl-3-hexanol 95%

Dodavatel: Apollo Scientific

OR931230-10G OR931230-1G OR931230-5G
APOSOR931230-10GEA 12830 CZK
APOSOR931230-10G APOSOR931230-1G APOSOR931230-5G
(±)-2,2-Dimethyl-3-hexanol 95%
(±)-2,2-Dimethyl-3-hexanol
Vzorec: C₈H₁₈O
MW: 130,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021929
CAS číslo: 4209-90-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR