4-Chloro-N,N-dimethylbutanamide 95%
Dodavatel: Apollo Scientific
OR931220-1G
OR931220-250MG
APOSOR931220-1GEA
12400
CZK
APOSOR931220-1G
APOSOR931220-250MG
4-Chloro-N,N-dimethylbutanamide 95%
4-Chloro-N,N-dimethylbutanamide
Vzorec:
C₆H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD04973966 CAS číslo: 22813-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...