Tisk…

n-Propyl carbamate 95%

Dodavatel: Apollo Scientific

Synonyms: Carbamic acid, propyl ester, O-propylcarbamate, Propyl urethane

OR931217-100MG OR931217-1G OR931217-250MG OR931217-5G OR931217-25G
APOSOR931217-100MGEA 0 CZK
APOSOR931217-100MG APOSOR931217-1G APOSOR931217-250MG APOSOR931217-5G APOSOR931217-25G
n-Propyl carbamate 95%
n-Propyl carbamate
Vzorec: C₄H₉NO₂
MW: 103,121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047879
CAS číslo: 627-12-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR