Tisk…

Citraconic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Methyl-2-butenedioic acid

OR931202-25G OR931202-100G OR931202-500G
APOSOR931202-25GEA 1490 CZK
APOSOR931202-25G APOSOR931202-100G APOSOR931202-500G
Citraconic acid 95%
Citraconic acid
Vzorec: C₅H₆O₄
MW: 130.1
Bod varu: 336
Storage Temperature: Ambient
MDL Number: MFCD00078086
CAS číslo: 498-23-7
Merck Index: 13,02345

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR