Tisk…

Benzotriazol-1-ylacetic acid 96%

Dodavatel: Apollo Scientific

Synonyms: 2-(1H-Benzo[d][1,2,3]triazol-1-yl)acetic acid

OR931189-1G
APOSOR931189-1GEA 5960 CZK
APOSOR931189-1G
Benzotriazol-1-ylacetic acid 96%
Benzotriazol-1-ylacetic acid
Vzorec: C₈H₇N₃O₂
MW: 177,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00553342
CAS číslo: 4144-64-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR