Tisk…

Isobutyl-4-hydroxybenzoate 96%

Dodavatel: Apollo Scientific

Synonyms: Isobutyl 4-hydroxybenzoate, Isobutylparaben

OR931176-100G OR931176-500G
APOSOR931176-100GEA 622 CZK
APOSOR931176-100G APOSOR931176-500G
Isobutyl-4-hydroxybenzoate 96%
Isobutyl-4-hydroxybenzoate
Vzorec: C₁₁H₁₄O₃
MW: 194,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020167
CAS číslo: 4247-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR