N-Chloroacetyl-DL-2-aminobutyric acid 95%
Dodavatel: Apollo Scientific
OR931113-25G
OR931113-5G
APOSOR931113-25GEA
9330
CZK
APOSOR931113-25G
APOSOR931113-5G
N-Chloroacetyl-DL-2-aminobutyric acid 95%
N-Chloroacetyl-DL-2-aminobutyric acid
Vzorec:
C₆H₁₀ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00055800 CAS číslo: 101072-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...