2,3-Dichloroquinoline 98%
Dodavatel: Apollo Scientific
OR931103-10G
OR931103-1G
OR931103-5G
APOSOR931103-10GEA
21100
CZK
APOSOR931103-10G
APOSOR931103-1G
APOSOR931103-5G
2,3-Dichloroquinoline 98%
2,3-Dichloroquinoline
Vzorec:
C₉H₅Cl₂N Storage Temperature: Ambient |
CAS číslo:
613-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...