Tisk…

4-Phenyl-5,6-dihydro-2H-pyran 97%

Dodavatel: Apollo Scientific

OR930655-1G OR930655-250MG
APOSOR930655-1GEA 20270 CZK
APOSOR930655-1G APOSOR930655-250MG
4-Phenyl-5,6-dihydro-2H-pyran 97%
4-Phenyl-5,6-dihydro-2H-pyran
Vzorec: C₁₁H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD18432773
CAS číslo: 3174-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR