4-Dimethylaminomethylthiophene-2-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR930506-1G
OR930506-5G
APOSOR930506-1GEA
4770
CZK
APOSOR930506-1G
APOSOR930506-5G
4-Dimethylaminomethylthiophene-2-carbaldehyde 95%
4-Dimethylaminomethylthiophene-2-carbaldehyde
Vzorec:
C₈H₁₁NOS MW: 169,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06803373 CAS číslo: 893745-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...