FOR-NLE-OH 95%
Dodavatel: Apollo Scientific
OR930495-1G
OR930495-250MG
APOSOR930495-1GEA
6220
CZK
APOSOR930495-1G
APOSOR930495-250MG
FOR-NLE-OH 95%
FOR-NLE-OH
Vzorec:
C₇H₁₃NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00237678 CAS číslo: 133388-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...