Tisk…

DL-Leucic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Hydroxy-4-methylpentanoic acid, Leucic acid, DL-2-hydroxy-4-methylvaleric acid, 2-Hydroxy-4-methylvaleric Acid

OR930492-100G OR930492-25G OR930492-5G
APOSOR930492-100GEA 10580 CZK
APOSOR930492-100G APOSOR930492-25G APOSOR930492-5G
DL-Leucic acid 95%
DL-Leucic acid
Vzorec: C₆H₁₂O₃
MW: 132,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004246
CAS číslo: 498-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR