Tisk…

6-Cyanopicolinic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR930215-250MG OR930215-1G
APOSOR930215-250MGEA 3710 CZK
APOSOR930215-250MG APOSOR930215-1G
6-Cyanopicolinic acid
6-Cyanopicolinic acid
Vzorec: C₇H₄N₂O₂
MW: 148,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10574696
CAS číslo: 872602-74-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR