4-Acetamido-2-methylbenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR9302-5G
OR9302-25G
APOSOR9302-5GEA
3230
CZK
APOSOR9302-5G
APOSOR9302-25G
4-Acetamido-2-methylbenzoic acid
4-Acetamido-2-methylbenzoic acid
Vzorec:
C₁₀H₁₁NO₃ MW: 193.21 g/mol Bod tání: 237…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD02258874 CAS číslo: 103204-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...