Methyl-2-hydroxy-3-phenylpropanoate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2-hydroxy-3-phenylpropanoate
OR930185-10G
OR930185-1G
OR930185-5G
APOSOR930185-10GEA
3310
CZK
APOSOR930185-10G
APOSOR930185-1G
APOSOR930185-5G
Methyl-2-hydroxy-3-phenylpropanoate 95%
Methyl-2-hydroxy-3-phenylpropanoate
Vzorec:
C₁₀H₁₂O₃ MW: 180,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06654233 CAS číslo: 13674-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...