3-[(Methylamino)methyl]phenol 95%
Dodavatel: Apollo Scientific
OR929900-1G
OR929900-5G
APOSOR929900-1GEA
3020
CZK
APOSOR929900-1G
APOSOR929900-5G
3-[(Methylamino)methyl]phenol 95%
3-[(Methylamino)methyl]phenol
Vzorec:
C₈H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD09733571 CAS číslo: 123926-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...