4-Methyl-6-phenylpyrimidin-2-amine 95%
Dodavatel: Apollo Scientific
OR929890-1G
OR929890-5G
APOSOR929890-1GEA
2860
CZK
APOSOR929890-1G
APOSOR929890-5G
4-Methyl-6-phenylpyrimidin-2-amine 95%
4-Methyl-6-phenylpyrimidin-2-amine
Vzorec:
C₁₁H₁₁N₃ Storage Temperature: Ambient |
MDL Number:
MFCD06123970 CAS číslo: 15755-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...