6-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
Dodavatel: Apollo Scientific
OR929870-1G
OR929870-250MG
APOSOR929870-1GEA
3810
CZK
APOSOR929870-1G
APOSOR929870-250MG
6-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
6-Methyl-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₈H₈N₂ MW: 132,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659664 CAS číslo: 824-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...