4-Methoxybenzensulphonic acid 95%
Dodavatel: Apollo Scientific
OR929851-1G
OR929851-25G
OR929851-5G
APOSOR929851-1GEA
2030
CZK
APOSOR929851-1G
APOSOR929851-25G
APOSOR929851-5G
4-Methoxybenzensulphonic acid 95%
4-Methoxybenzensulphonic acid
Vzorec:
C₇H₈O₄S Storage Temperature: Ambient |
MDL Number:
MFCD08685124 CAS číslo: 5857-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...