Tisk…

3-Chloro-4-propoxybenzaldehyde 95%

Dodavatel: Apollo Scientific

OR929821-1G OR929821-5G
APOSOR929821-1GEA 6220 CZK
APOSOR929821-1G APOSOR929821-5G
3-Chloro-4-propoxybenzaldehyde 95%
3-Chloro-4-propoxybenzaldehyde
Vzorec: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246436
CAS číslo: 99070-71-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR