Tisk…

4-Chloro-2-methyl-6-(methylthio)pyrimidine 95%

Dodavatel: Apollo Scientific

OR929794-1G
APOSOR929794-1GEA 4630 CZK
APOSOR929794-1G
4-Chloro-2-methyl-6-(methylthio)pyrimidine 95%
4-Chloro-2-methyl-6-(methylthio)pyrimidine
Vzorec: C₆H₇ClN₂S
Storage Temperature: Ambient
MDL Number: MFCD16769133
CAS číslo: 867131-59-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR