Tisk…

4-Chloro-5-methoxypyrimidine 95%

Dodavatel: Apollo Scientific

OR929762-1G
APOSOR929762-1GEA 2730 CZK
APOSOR929762-1G
4-Chloro-5-methoxypyrimidine 95%
4-Chloro-5-methoxypyrimidine
Vzorec: C₅H₅ClN₂O
MW: 144,5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09966063
CAS číslo: 695-85-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR