2-Nitrocyclohexanone 95%
Dodavatel: Apollo Scientific
OR929759-1G
OR929759-250MG
APOSOR929759-1GEA
10350
CZK
APOSOR929759-1G
APOSOR929759-250MG
2-Nitrocyclohexanone 95%
2-Nitrocyclohexanone
Vzorec:
C₆H₉NO₃ Storage Temperature: Ambient |
CAS číslo:
4883-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...