Tisk…

(Z)-3-Bromopropenoic acid methyl ester 95/5 Z/E 95%

Dodavatel: Apollo Scientific

OR929700-1G OR929700-250MG
APOSOR929700-1GEA 9330 CZK
APOSOR929700-1G APOSOR929700-250MG
(Z)-3-Bromopropenoic acid methyl ester 95/5 Z/E 95%
(Z)-3-Bromopropenoic acid methyl ester 95/5 Z/E
Vzorec: C₄H₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11109369
CAS číslo: 6214-22-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR