Tisk…

2,4-Dimethylphenylacetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: (2,4-Dimethylphenyl)acetic acid, 2,4-Xylylacetic acid

OR929615-100G OR929615-10G OR929615-25G
APOSOR929615-100GEA 17130 CZK
APOSOR929615-100G APOSOR929615-10G APOSOR929615-25G
2,4-Dimethylphenylacetic acid 98%
2,4-Dimethylphenylacetic acid
Vzorec: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664685
CAS číslo: 6331-04-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR