Tisk…

7-Thieno[3,2-d]pyrimidine 95%

Dodavatel: Apollo Scientific

OR929592-1G OR929592-250MG
APOSOR929592-1GEA 12870 CZK
APOSOR929592-1G APOSOR929592-250MG
7-Thieno[3,2-d]pyrimidine 95%
7-Thieno[3,2-d]pyrimidine
Vzorec: C₆H₄N₂S
Storage Temperature: Ambient
MDL Number: MFCD11110439
CAS číslo: 272-68-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR