Tisk…

(2,4-Dichlorothiazol-5-yl)methanol 95%

Dodavatel: Apollo Scientific

OR929547-1G OR929547-250MG
APOSOR929547-1GEA 14880 CZK
APOSOR929547-1G APOSOR929547-250MG
(2,4-Dichlorothiazol-5-yl)methanol 95%
(2,4-Dichlorothiazol-5-yl)methanol
Vzorec: C₄H₃Cl₂NOS
Storage Temperature: Ambient
CAS číslo: 170232-69-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR