Tisk…

4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 98%

Dodavatel: Apollo Scientific

Synonyms: 1,2,2,6,6-Pentamethylpiperidin-4-ol, 1,2,2,6,6-Pentamethyl-4-piperidinol

OR929421-500G
APOSOR929421-500GEA 2400 CZK
APOSOR929421-500G
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine 98%
4-Hydroxy-1,2,2,6,6-pentamethylpiperidine
Vzorec: C₁₀H₂₁NO
MW: 171,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191220
CAS číslo: 2403-89-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR