Tisk…

2-Ethyl-3-methoxybenzoic acid 95%

Dodavatel: Apollo Scientific

OR929404-5G
APOSOR929404-5GEA 2440 CZK
APOSOR929404-5G
2-Ethyl-3-methoxybenzoic acid 95%
2-Ethyl-3-methoxybenzoic acid
Vzorec: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11100772
CAS číslo: 57598-51-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR