Tisk…

2-Isopropylbenzoic acid 98%

Dodavatel: Apollo Scientific

OR929391-1G OR929391-5G
APOSOR929391-1GEA 3380 CZK
APOSOR929391-1G APOSOR929391-5G
2-Isopropylbenzoic acid 98%
2-Isopropylbenzoic acid
Vzorec: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00045836
CAS číslo: 2438-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR