Tisk…

Fmoc-HomoArg(Pbf)-OH 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulphonyl)guanidino)hexanoic acid, Fmoc-HoArg(Pbf)-OH

OR929330-10G OR929330-25G OR929330-5G
APOSOR929330-10GEA 6290 CZK
APOSOR929330-10G APOSOR929330-25G APOSOR929330-5G
Fmoc-HomoArg(Pbf)-OH 95%
Fmoc-HomoArg(Pbf)-OH
Vzorec: C₃₅H₄₂N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD03093571
CAS číslo: 1159680-21-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR