Ethyl-3-oxocyclopentane-1-carboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 3-oxocyclopentanecarboxylate
OR929246-5G
APOSOR929246-5GEA
1700
CZK
APOSOR929246-5G
Ethyl-3-oxocyclopentane-1-carboxylate 97%
Ethyl-3-oxocyclopentane-1-carboxylate
Vzorec:
C₈H₁₂O₃ MW: 156,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01320352 CAS číslo: 5400-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...