Tisk…

3-Chloro-4-nitrophenol 98%

Dodavatel: Apollo Scientific

OR929226-10G
APOSOR929226-10GEA 2480 CZK
APOSOR929226-10G
3-Chloro-4-nitrophenol 98%
3-Chloro-4-nitrophenol
Vzorec: C₆H₄ClNO₃
MW: 173,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466090
CAS číslo: 491-11-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR