Tisk…
(2-Ethoxyphenyl)acetic acid 95%
Dodavatel: Apollo Scientific
Synonyms: 2-Ethoxyphenylacetic acid, 2-(2-Ethoxyphenyl)acetic acid
OR929038-5G
APOSOR929038-5GEA 3190 CZK
APOSOR929038-5G
(2-Ethoxyphenyl)acetic acid 95%
(2-Ethoxyphenyl)acetic acid
Vzorec: H₃CCH₂O(C₆H₄)CH₂COOH
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016824
CAS číslo: 70289-12-2