4-Mercaptophenylacetic acid 95%
Dodavatel: Apollo Scientific
OR928932-1G
OR928932-5G
APOSOR928932-1GEA
1870
CZK
APOSOR928932-1G
APOSOR928932-5G
4-Mercaptophenylacetic acid 95%
4-Mercaptophenylacetic acid
Vzorec:
C₈H₈O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00797617 CAS číslo: 39161-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...