Methyl-2-bromo-5-(bromomethyl)benzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-bromo-5-(bromomethyl)benzoate
OR928929-1G
OR928929-250MG
OR928929-5G
APOSOR928929-1GEA
8900
CZK
APOSOR928929-1G
APOSOR928929-250MG
APOSOR928929-5G
Methyl-2-bromo-5-(bromomethyl)benzoate 95%
Methyl-2-bromo-5-(bromomethyl)benzoate
Vzorec:
C₉H₈Br₂O₂ MW: 307.9666 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18398708 CAS číslo: 90721-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...