Ethyl-2-amino-1-cyclopentene-1-carboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 2-aminocyclopent-1-enecarboxylate
OR928805-1G
OR928805-25G
OR928805-5G
APOSOR928805-1GEA
2650
CZK
APOSOR928805-1G
APOSOR928805-25G
APOSOR928805-5G
Ethyl-2-amino-1-cyclopentene-1-carboxylate 95%
Ethyl-2-amino-1-cyclopentene-1-carboxylate
Vzorec:
C₈H₁₃NO₂ MW: 155,197 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00216929 CAS číslo: 7149-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...