Tisk…

Sarcosine ethyl ester hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride

OR928751-500G
APOSOR928751-500GEA 6620 CZK
APOSOR928751-500G
Sarcosine ethyl ester hydrochloride 97%
Sarcosine ethyl ester hydrochloride
Vzorec: C₅H₁₁NO₂·HCl
MW: 153,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012506
CAS číslo: 52605-49-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR