Tisk…

trans-2-Phenylcyclohexanol 95%

Dodavatel: Apollo Scientific

Synonyms: trans-2-Phenyl-1-cyclohexanol

OR928729-1G OR928729-5G
APOSOR928729-1GEA 6020 CZK
APOSOR928729-1G APOSOR928729-5G
trans-2-Phenylcyclohexanol 95%
trans-2-Phenylcyclohexanol
Vzorec: C₁₂H₁₆O
MDL Number: MFCD00003865
CAS číslo: 2362-61-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR