2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR928689-1G
OR928689-5G
APOSOR928689-1GEA
3940
CZK
APOSOR928689-1G
APOSOR928689-5G
2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde 95%
2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde
Vzorec:
C₁₀H₁₅NO Storage Temperature: Ambient |
MDL Number:
MFCD04624247 CAS číslo: 18870-75-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...