Tisk…

Tetrahydro-2H-thiopyran-4-ol 98%

Dodavatel: Apollo Scientific

OR928607-10G OR928607-1G OR928607-5G
APOSOR928607-10GEA 15840 CZK
APOSOR928607-10G APOSOR928607-1G APOSOR928607-5G
Tetrahydro-2H-thiopyran-4-ol 98%
Tetrahydro-2H-thiopyran-4-ol
Vzorec: C₅H₁₀OS
MW: 118,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02942489
CAS číslo: 29683-23-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR