Tisk…

2-Methyl-2-propenyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: Acetic Acid 2-Methyl-2-propenyl Ester, Acetic Acid 2-Methylallyl Ester, beta-Methallyl Acetate, 2-Methylallyl Acetate

OR928563-25G
APOSOR928563-25GEA 3190 CZK
APOSOR928563-25G
2-Methyl-2-propenyl acetate 95%
2-Methyl-2-propenyl acetate
Vzorec: C₆H₁₀O₂
MW: 114,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048215
CAS číslo: 820-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR